Boc-(2S,3S)-3-amino-2-hydroxy-5-methylhexanoic acid
Supplier: Matrix Scientific
042071-100MG
042071-500MG
042071-1G
101854-338EA
275.38
USD
101854-338
101854-340
101854-342
Boc-(2S,3S)-3-amino-2-hydroxy-5-methylhexanoic acid
Boc-(2S,3S)-3-amino-2-hydroxy-5-methylhexanoic acid
Hazard: Irritant
Formula:
C₁₂H₂₃NO₅ MW: 261.32 g/mol |
MDL Number:
MFCD04974450 CAS Number: 73397-27-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...