Boc-(2S,3S)-2-amino-3-methoxybutanoic acid
Supplier: Matrix Scientific
042070-100MG
042070-250MG
101854-334EA
630.77
USD
101854-334
101854-336
Boc-(2S,3S)-2-amino-3-methoxybutanoic acid
Boc-(2S,3S)-2-amino-3-methoxybutanoic acid
Hazard: Irritant
Formula:
C₁₀H₁₉NO₅ |
MDL Number:
MFCD02682599 CAS Number: 630424-73-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...