Boc-(2S,3R)-3-amino-2-hydroxy-5-methylhexanoic acid

Supplier: Matrix Scientific

042067-100MG 042067-500MG 042067-1G
101854-320EA 275.38 USD
101854-320 101854-322 101854-324
Boc-(2S,3R)-3-amino-2-hydroxy-5-methylhexanoic acid
Boc-(2S,3R)-3-amino-2-hydroxy-5-methylhexanoic acid

Hazard: Irritant

Formula: C₁₂H₂₃NO₅
MDL Number: MFCD00070262
CAS Number: 73397-25-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR