Boc-(2R,3S)-3-amino-2-hydroxy-5-methylhexanoic acid
Supplier: Matrix Scientific
042066-100MG
042066-500MG
042066-1G
101854-314EA
275.38
USD
101854-314
101854-316
101854-318
Boc-(2R,3S)-3-amino-2-hydroxy-5-methylhexanoic acid
Boc-(2R,3S)-3-amino-2-hydroxy-5-methylhexanoic acid
Hazard: Irritant
Formula:
C₁₂H₂₃NO₅ |
MDL Number:
MFCD04972275 CAS Number: 73397-28-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...