Boc-(2R,3R)-3-amino-2-hydroxy-5-methylhexanoic acid

Supplier: Matrix Scientific

042064-100MG 042064-500MG 042064-1G
101854-304EA 275.38 USD
101854-304 101854-306 101854-308
Boc-(2R,3R)-3-amino-2-hydroxy-5-methylhexanoic acid
Boc-(2R,3R)-3-amino-2-hydroxy-5-methylhexanoic acid

Hazard: Irritant

Formula: C₁₂H₂₃NO₅
MDL Number: MFCD00058525
CAS Number: 73397-26-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR