Fmoc-trans-4-hydroxy-D-proline
Supplier: Matrix Scientific
Synonyms:
Fmoc-trans-D-Hyp-OH, (2R,4S)-1-(((9H-Fluoren-9-yl)methoxy)carbonyl)-4-hydroxypyrrolidine-2-carboxylic acid
042063-500MG
042063-1G
101854-300EA
596.59
USD
101854-300
101854-302
Fmoc-trans-4-hydroxy-D-proline
Fmoc-trans-4-hydroxy-D-proline
Hazard: Irritant
Formula:
C₂₀H₁₉NO₅ MW: 353.38 g/mol |
MDL Number:
MFCD01862677 CAS Number: 139262-20-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...