Fmoc-trans-4-fluoro-Pro-Oh

Supplier: Matrix Scientific

Synonyms: (2S,4R)-1-(((9H-Fluoren-9-yl)methoxy)carbonyl)-4-fluoropyrrolidine-2-carboxylic acid, (2S,4R)-4-Fluoropyrrolidine-2-carboxylic acid, N-FMOC protected, Fmoc-trans-4-F-Pro-OH

042062-500MG 042062-1G
101854-296EA 958.46 USD
101854-296 101854-298
Fmoc-trans-4-fluoro-Pro-Oh
Fmoc-trans-4-fluoro-Pro-Oh

Hazard: Irritant

Formula: C₂₀H₁₈FNO₄
MDL Number: MFCD03424224
CAS Number: 203866-20-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR