Fmoc-D-CIS-HYP-OH

Supplier: Matrix Scientific

042061-500MG 042061-1G
101854-292EA 760 USD
101854-292 101854-294
Fmoc-D-CIS-HYP-OH
Fmoc-D-CIS-HYP-OH

Hazard: Irritant

Formula: C₂₀H₁₉NO₅
MDL Number: MFCD01318735
CAS Number: 214852-45-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR