AmbotzFAA1495

Supplier: Matrix Scientific

Synonyms: (S)-3-(((9H-Fluoren-9-yl)methoxy)carbonyl)thiazolidine-4-carboxylic acid, Fmoc-D-thiazolidine-4-carboxylic acid

042058-1G 042058-5G
101854-280EA 224.62 USD
101854-280 101854-282
AmbotzFAA1495
AmbotzFAA1495

Hazard: Irritant

Formula: C₁₉H₁₇NO₄S
MW: 355.408 g/mol
MDL Number: MFCD00235911
CAS Number: 198545-89-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR