(S)-N-Fmoc-azetidine-2-carboxylic acid
Supplier: Matrix Scientific
Synonyms:
(S)-1-(((9H-Fluoren-9-yl)methoxy)carbonyl)azetidine-2-carboxylic acid, Fmoc-L-azetidine-2-carboxylic acid
042056-1G
042056-5G
042056-10G
101854-270EA
363.08
USD
101854-270
101854-272
101854-274
(S)-N-Fmoc-azetidine-2-carboxylic acid
(S)-N-Fmoc-azetidine-2-carboxylic acid
Hazard: Irritant
Formula:
C₁₉H₁₇NO₄ MW: 323.348 g/mol |
MDL Number:
MFCD01320843 CAS Number: 136552-06-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...