Fmoc-Thz-OH

Supplier: Matrix Scientific

042055-1G 042055-5G
101854-266EA 113.85 USD
101854-266 101854-268
Fmoc-Thz-OH

Hazard: Irritant

Formula: C₁₉H₁₇NO₄S
MDL Number: MFCD00235910
CAS Number: 133054-21-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR