Fmoc-(R)-thiazolidine-2-carboxylic acid

Supplier: Matrix Scientific

042054-1G 042054-5G
101854-262EA 412.31 USD
101854-262 101854-264
Fmoc-(R)-thiazolidine-2-carboxylic acid
Fmoc-(R)-thiazolidine-2-carboxylic acid

Hazard: Irritant

Formula: C₁₉H₁₇NO₄S
MDL Number: MFCD02682345
CAS Number: 1217481-34-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR