Fmoc-(4S,2RS)-2-phenylthiazolidine-4-carboxylic acid

Supplier: Matrix Scientific

042052-1G 042052-5G
101854-254EA 635.38 USD
101854-254 101854-256
Fmoc-(4S,2RS)-2-phenylthiazolidine-4-carboxylic acid
Fmoc-(4S,2RS)-2-phenylthiazolidine-4-carboxylic acid

Hazard: Irritant

Formula: C₂₅H₂₁NO₄S
MDL Number: MFCD02682339
CAS Number: 1217510-53-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR