Fmoc-(4S,2RS)-2-methylthiazolidine-4-carboxylicacid

Supplier: Matrix Scientific

042051-1G 042051-5G
101854-250EA 598.46 USD
101854-250 101854-252
Fmoc-(4S,2RS)-2-methylthiazolidine-4-carboxylicacid
Fmoc-(4S,2RS)-2-methylthiazolidine-4-carboxylicacid

Hazard: Irritant

Formula: C₂₀H₁₉NO₄S
MDL Number: MFCD02682332
CAS Number: 1217544-28-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR