Fmoc-(4S,2RS)-2-ethylthiazolidine-4-carboxylicacid

Supplier: Matrix Scientific

042050-500MG 042050-1G
101854-246EA 412.31 USD
101854-246 101854-248
Fmoc-(4S,2RS)-2-ethylthiazolidine-4-carboxylicacid
Fmoc-(4S,2RS)-2-ethylthiazolidine-4-carboxylicacid

Hazard: Irritant

Formula: C₂₁H₂₁NO₄S
MDL Number: MFCD02682323
CAS Number: 1217512-44-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR