Fmoc-(2S,4S)-4-phenylpyrrolidine-2-carboxylic acid

Supplier: Matrix Scientific

Synonyms: (2S,4S)-1-(((9H-Fluoren-9-yl)methoxy)carbonyl)-4-phenylpyrrolidine-2-carboxylic acid

042048-250MG 042048-1G
101854-238EA 412.31 USD
101854-238 101854-240
Fmoc-(2S,4S)-4-phenylpyrrolidine-2-carboxylic acid
Fmoc-(2S,4S)-4-phenylpyrrolidine-2-carboxylic acid

Hazard: Irritant

Formula: C₂₆H₂₃NO₄
MW: 413.48 g/mol
MDL Number: MFCD01074697
CAS Number: 269078-71-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR