Fmoc-(2S,4S)-(-)-4-hydroxypyrrolidine-2-carboxylic acid
Supplier: Matrix Scientific
Synonyms:
(2S,4S)-1-(((9H-Fluoren-9-yl)methoxy)carbonyl)-4-hydroxypyrrolidine-2-carboxylic acid
042047-250MG
042047-1G
101854-234EA
275.38
USD
101854-234
101854-236
Fmoc-(2S,4S)-(-)-4-hydroxypyrrolidine-2-carboxylic acid
Fmoc-(2S,4S)-(-)-4-hydroxypyrrolidine-2-carboxylic acid
Hazard: Irritant
Formula:
C₂₀H₁₉NO₅ MW: 353.38 g/mol |
MDL Number:
MFCD02682328 CAS Number: 189249-10-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...