Fmoc-Hyp-OH

Supplier: Matrix Scientific

Synonyms: N-Fmoc-trans-4-hydroxy-L-proline

042045-5G 042045-25G 042045-100G
101854-228EA 53.85 USD
101854-228 102663-706 102663-708
Fmoc-Hyp-OH
Fmoc-Hyp-OH

Hazard: Irritant

Formula: C₂₀H₁₉NO₅
MW: 353.374 g/mol
MDL Number: MFCD00151929
CAS Number: 88050-17-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR