Fmoc-(2S,3S)-3-hydroxypyrrolidine-2-carboxylicacid

Supplier: Matrix Scientific

Synonyms: Fmoc-trans-3-OH-Pro-OH, (2S,3S)-1-(((9H-Fluoren-9-yl)methoxy)carbonyl)-3-hydroxypyrrolidine-2-carboxylic acid

042043-1G 042043-5G
101854-220EA 1376.92 USD
101854-220 101854-222
Fmoc-(2S,3S)-3-hydroxypyrrolidine-2-carboxylicacid
Fmoc-(2S,3S)-3-hydroxypyrrolidine-2-carboxylicacid

Hazard: Irritant

Formula: C₂₀H₁₉NO₅
MDL Number: MFCD02094543
CAS Number: 296774-32-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR