Fmoc-Oic-OH

Supplier: Matrix Scientific

Synonyms: (2S,3aS,7aS)-1-Fmoc-octahydroindole-2-carboxylic acid, (2S,3aS,7aS)-1-(((9H-Fluoren-9-yl)methoxy)carbonyl)octahydro-1H-indole-2-carboxylic acid

042041-1G 042041-5G
101854-212EA 709.23 USD
101854-212 101854-214
Fmoc-Oic-OH
Fmoc-Oic-OH

Hazard: Irritant

Formula: C₂₄H₂₅NO₄
MDL Number: MFCD00191201
CAS Number: 130309-37-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR