Fmoc-(2S,4R)-4-benzylpyrrolidine-2-carboxylicacid

Supplier: Matrix Scientific

Synonyms: (2S,4R)-1-(((9H-Fluoren-9-yl)methoxy)carbonyl)-4-benzylpyrrolidine-2-carboxylic acid

042040-500MG 042040-1G
101854-208EA 375.38 USD
101854-208 101854-210
Fmoc-(2S,4R)-4-benzylpyrrolidine-2-carboxylicacid
Fmoc-(2S,4R)-4-benzylpyrrolidine-2-carboxylicacid

Hazard: Irritant

Formula: C₂₇H₂₅NO₄
MW: 427.5 g/mol
MDL Number: MFCD04115782
CAS Number: 1158891-05-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR