Fmoc-(2R,5S)-5-phenylpyrrolidine-2-carboxylicacid
Supplier: Matrix Scientific
042039-250MG
042039-1G
101854-204EA
461.54
USD
101854-204
101854-206
Fmoc-(2R,5S)-5-phenylpyrrolidine-2-carboxylicacid
Fmoc-(2R,5S)-5-phenylpyrrolidine-2-carboxylicacid
Hazard: Irritant
Formula:
C₂₆H₂₃NO₄ |
MDL Number:
MFCD01074695 CAS Number: 269078-69-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...