(2S,4R)-1-(tert-Butoxycarbonyl)-4-fluoro-2-pyrrolidinecarboxylic acid

Supplier: Matrix Scientific

Synonyms: N-(tert-Butoxycarbonyl)-trans-4-fluoro-L-proline, (2S,4R)-1-Boc-4-fluoro-2-pyrrolidinecarboxylic Acid, (2S,4R)-1-(tert-Butoxycarbonyl)-4-fluoropyrrolidine-2-carboxylic acid, N-Boc-trans-4-fluoro-L-proline

042037-500MG 042037-1G
101854-196EA 272.31 USD
101854-196 101854-198
(2S,4R)-1-(tert-Butoxycarbonyl)-4-fluoro-2-pyrrolidinecarboxylic acid
(2S,4R)-1-(tert-Butoxycarbonyl)-4-fluoro-2-pyrrolidinecarboxylic acid

Hazard: Irritant

Formula: C₁₀H₁₆FNO₄
MW: 233.24 g/mol
MDL Number: MFCD06796085
CAS Number: 203866-14-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR