Boc-trans-4-amino-L-proline methyl ester hydrochloride salt
Supplier: Matrix Scientific
Synonyms:
(2S,4R)-1-tert-Butyl 2-methyl 4-aminopyrrolidine-1,2-dicarboxylate hydrochloride, Methyl (2S,4R)-4-aminopyrrolidine-2-carboxylate hydrochloride, N1-BOC protected
042035-500MG
042035-1G
101854-188EA
369.23
USD
101854-188
101854-190
Boc-trans-4-amino-L-proline methyl ester hydrochloride salt
Boc-trans-4-amino-L-proline methyl ester hydrochloride salt
Hazard: Irritant
Formula:
C₁₁H₂₁ClN₂O₄ MW: 280.75 g/mol |
MDL Number:
MFCD03787929 CAS Number: 334999-32-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...