cis-N-(tert-Butoxycarbonyl)-4-hydroxy-D-proline

Supplier: Matrix Scientific

Synonyms: Boc-cis-D-Hyp-OH, (2R,4R)-1-(tert-Butoxycarbonyl)-4-hydroxypyrrolidine-2-carboxylic acid, (2R,4R)-N-Boc-4-hydroxy-2-pyrrolidinecarboxylic acid

042034-1G 042034-5G
101854-186EA 20 USD
101854-186 102551-300
cis-N-(tert-Butoxycarbonyl)-4-hydroxy-D-proline
cis-N-(tert-Butoxycarbonyl)-4-hydroxy-D-proline

Hazard: Irritant

Formula: C₁₀H₁₇NO₅
MW: 231.25 g/mol
Melting Pt: ∼ 145 °C (dec.)
MDL Number: MFCD02094407
CAS Number: 135042-12-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR