cis-N-Boc-4-cyano-L-proline methyl ester

Supplier: Matrix Scientific

Synonyms: Boc-cis-4-CN-Pro-OMe, (2S,4S)-1-tert-Butyl 2-methyl 4-cyanopyrrolidine-1,2-dicarboxylate, 1-tert-Butyl 2-methyl (2S,4S)-4-cyanopyrrolidine-1,2-dicarboxylate

042032-500MG 042032-1G
101854-176EA 370.77 USD
101854-176 101854-178
cis-N-Boc-4-cyano-L-proline methyl ester
cis-N-Boc-4-cyano-L-proline methyl ester

Hazard: Irritant

Formula: C₁₂H₁₈N₂O₄
MDL Number: MFCD03787928
CAS Number: 487048-28-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR