Boc-cis-4-amino-L-proline methyl ester hydrochloride salt

Supplier: Matrix Scientific

Synonyms: Methyl (2S,4S)-1-Boc-4-aminopyrrolidine-2-carboxylate hydrochloride

042031-500MG 042031-1G
101854-172EA 149.23 USD
101854-172 101854-174
Boc-cis-4-amino-L-proline methyl ester hydrochloride salt
Boc-cis-4-amino-L-proline methyl ester hydrochloride salt

Hazard: Irritant

Formula: C₁₁H₂₁ClN₂O₄
MW: 280.75 g/mol
MDL Number: MFCD03787927
CAS Number: 171110-72-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR