Boc-3,4-dehydro-L-proline

Supplier: Matrix Scientific

Synonyms: (S)-1-(tert-Butoxycarbonyl)-2,5-dihydro-1H-pyrrole-2-carboxylic acid, Boc-3,4-dehydro-Pro-OH

042030-1G 042030-500MG
101854-168EA 686.15 USD
101854-168 101854-170
Boc-3,4-dehydro-L-proline
Boc-3,4-dehydro-L-proline

Hazard: Irritant

Formula: C₁₀H₁₅NO₄
MW: 213.233 g/mol
MDL Number: MFCD00037896
CAS Number: 51154-06-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR