Boc-(S)-thiazolidine-2-carboxylic acid

Supplier: Matrix Scientific

Synonyms: (S)-3-(tert-Butoxycarbonyl)thiazolidine-2-carboxylic acid

042028-1G 042028-5G
101854-160EA 412.31 USD
101854-160 101854-162
Boc-(S)-thiazolidine-2-carboxylic acid
Boc-(S)-thiazolidine-2-carboxylic acid

Hazard: Irritant

Formula: C₉H₁₅NO₄S
MW: 233.29 g/mol
MDL Number: MFCD02682342
CAS Number: 891192-95-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR