Boc-(R)-thiazolidine-2-carboxylic acid

Supplier: Matrix Scientific

042025-1G 042025-5G
101854-148EA 412.31 USD
101854-148 101854-150
Boc-(R)-thiazolidine-2-carboxylic acid
Boc-(R)-thiazolidine-2-carboxylic acid

Hazard: Irritant

Formula: C₉H₁₅NO₄S
MDL Number: MFCD02682344
CAS Number: 125471-00-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR