Boc-(4S,2RS)-2-phenylthiazolidine-4-carboxylicacid

Supplier: Matrix Scientific

042023-1G 042023-5G
101854-140EA 635.38 USD
101854-140 101854-142
Boc-(4S,2RS)-2-phenylthiazolidine-4-carboxylicacid
Boc-(4S,2RS)-2-phenylthiazolidine-4-carboxylicacid

Hazard: Irritant

Formula: C₁₅H₁₉NO₄S
MDL Number: MFCD02682338
CAS Number: 637032-03-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR