Boc-(4S,2RS)-2-methylthiazolidine-4-carboxylicacid

Supplier: Matrix Scientific

042022-1G 042022-5G
101854-136EA 598.46 USD
101854-136 101854-138
Boc-(4S,2RS)-2-methylthiazolidine-4-carboxylicacid
Boc-(4S,2RS)-2-methylthiazolidine-4-carboxylicacid

Hazard: Irritant

Formula: C₁₀H₁₇NO₄S
MDL Number: MFCD02682331
CAS Number: 1217546-87-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR