Boc-(4S,2RS)-2-ethylthiazolidine-4-carboxylic acid

Supplier: Matrix Scientific

042021-500MG 042021-1G
101854-132EA 412.31 USD
101854-132 101854-134
Boc-(4S,2RS)-2-ethylthiazolidine-4-carboxylic acid
Boc-(4S,2RS)-2-ethylthiazolidine-4-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₉NO₄S
MDL Number: MFCD02682322
CAS Number: 1217515-06-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR