Boc-(2S,5R)-5-phenylpyrrolidine-2-carboxylic acid
Supplier: Matrix Scientific
Synonyms:
(2S,5R)-1-(tert-Butoxycarbonyl)-5-phenylpyrrolidine-2-carboxylic acid
042020-250MG
042020-1G
101854-128EA
238.46
USD
101854-128
101854-130
Boc-(2S,5R)-5-phenylpyrrolidine-2-carboxylic acid
Boc-(2S,5R)-5-phenylpyrrolidine-2-carboxylic acid
Hazard: Irritant
Formula:
C₁₆H₂₁NO₄ MW: 291.35 g/mol |
MDL Number:
MFCD01321008 CAS Number: 221352-49-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...