Boc-(2S,4S)-4-phenylpyrrolidine-2-carboxylic acid
Supplier: Matrix Scientific
Synonyms:
(2S,4S)-1-(tert-Butoxycarbonyl)-4-phenylpyrrolidine-2-carboxylic acid
042019-250MG
042019-1G
101854-124EA
412.31
USD
101854-124
101854-126
Boc-(2S,4S)-4-phenylpyrrolidine-2-carboxylic acid
Boc-(2S,4S)-4-phenylpyrrolidine-2-carboxylic acid
Hazard: Irritant
Formula:
C₁₆H₂₁NO₄ |
MDL Number:
MFCD01321010 CAS Number: 96314-29-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...