Boc-trans-3-hydroxy-L-proline
Supplier: Matrix Scientific
Synonyms:
(2S,3S)-1-(tert-Butoxycarbonyl)-3-hydroxypyrrolidine-2-carboxylic acid, Boc-trans-3-OH-Pro-OH
042016-500MG
042016-1G
042016-5G
101854-110EA
915.38
USD
101854-110
101854-112
101854-114
Boc-trans-3-hydroxy-L-proline
Boc-trans-3-hydroxy-L-proline
Hazard: Irritant
Formula:
C₁₀H₁₇NO₅ MW: 231.25 g/mol |
MDL Number:
MFCD04974546 CAS Number: 187039-57-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...