Boc-L-octahydroindole-2-carboxylic acid

Supplier: Matrix Scientific

Synonyms: Boc-Oic-OH, (2S,3aS,7aS)-1-(tert-Butoxycarbonyl)octahydro-1H-indole-2-carboxylic acid, Boc-(2S,3aS,7aS)-octahydro-1H-indole-2-carboxylic acid

042014-1G 042014-5G
101854-102EA 367.69 USD
101854-102 101854-104
Boc-L-octahydroindole-2-carboxylic acid
Boc-L-octahydroindole-2-carboxylic acid

Hazard: Irritant

Formula: C₁₄H₂₃NO₄
MDL Number: MFCD09750486
CAS Number: 109523-13-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR