Boc-(2S,4R)-4-benzylpyrrolidine-2-carboxylic acid

Supplier: Matrix Scientific

Synonyms: (2S,4R)-4-Benzyl-1-(tert-butoxycarbonyl)pyrrolidine-2-carboxylic acid

042013-500MG 042013-1G
101854-098EA 532.31 USD
101854-098 101854-100
Boc-(2S,4R)-4-benzylpyrrolidine-2-carboxylic acid
Boc-(2S,4R)-4-benzylpyrrolidine-2-carboxylic acid

Hazard: Irritant

Formula: C₁₇H₂₃NO₄
MW: 305.38 g/mol
MDL Number: MFCD04115780
CAS Number: 153074-95-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR