Boc-(2R,5S)-5-phenylpyrrolidine-2-carboxylic acid

Supplier: Matrix Scientific

042012-250MG 042012-1G
101854-094EA 461.54 USD
101854-094 101854-096
Boc-(2R,5S)-5-phenylpyrrolidine-2-carboxylic acid
Boc-(2R,5S)-5-phenylpyrrolidine-2-carboxylic acid

Hazard: Irritant

Formula: C₁₆H₂₁NO₄
MDL Number: MFCD01321009
CAS Number: 158706-46-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR