Fmoc-L-Tyr(Me)-OH

Supplier: Matrix Scientific

Synonyms: Fmoc-Tyr(Me)-OH, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-O-methyl-L-tyrosine, N-Fmoc-4-methoxy-L-phenylalanine, N-Fmoc-O-methyl-L-tyrosine, Fmoc-Phe(4-OMe)-OH, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-4-methoxy-L-phenylalanine

042011-1G 042011-5G
101854-090EA 326.15 USD
101854-090 101854-092
Fmoc-L-Tyr(Me)-OH
Fmoc-L-Tyr(Me)-OH

Hazard: Irritant

Formula: C₂₅H₂₃NO₅
MW: 417.47 g/mol
MDL Number: MFCD00153368
CAS Number: 77128-72-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR