Fmoc-O-ethyl-D-tyrosine

Supplier: Matrix Scientific

Synonyms: Fmoc-D-Tyr(4-Et)-OH, Fmoc-D-Tyr(Et)-OH

042008-1G 042008-5G
101854-078EA 81.54 USD
101854-078 101854-080
Fmoc-O-ethyl-D-tyrosine
Fmoc-O-ethyl-D-tyrosine

Hazard: Irritant

Formula: C₂₆H₂₅NO₅
MW: 431.488 g/mol
MDL Number: MFCD00237035
CAS Number: 162502-65-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR