Fmoc-L-thyroxine
Supplier: Matrix Scientific
Synonyms:
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl)propanoic acid, Fmoc-Thyroxine
042007-1G
042007-5G
101854-074EA
686.15
USD
101854-074
101854-076
Fmoc-L-thyroxine
Fmoc-L-thyroxine
Hazard: Irritant
Formula:
C₃₀H₂₁I₄NO₆ |
MDL Number:
MFCD01861316 CAS Number: 151889-56-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...