Fmoc-L-thyronine

Supplier: Matrix Scientific

042006-1G 042006-5G
101854-070EA 1095.38 USD
101854-070 101854-072
Fmoc-L-thyronine
Fmoc-L-thyronine

Hazard: Irritant

Formula: C₃₀H₂₅NO₆
MDL Number: MFCD01861315
CAS Number: 1354485-04-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR