Fmoc-L-3,3-diphenylalanine ≥95%

Supplier: Matrix Scientific

042004-1G 042004-5G
101854-062EA 113.85 USD
101854-062 101854-064
Fmoc-L-3,3-diphenylalanine ≥95%
Fmoc-L-3,3-diphenylalanine

Hazard: Irritant

Formula: C₃₀H₂₅NO₄
MDL Number: MFCD00672337
CAS Number: 201484-50-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR