Fmoc-L-(2,6-di-Me)Tyr-OH

Supplier: Matrix Scientific

042003-250MG 042003-1G
101854-058EA 846.15 USD
101854-058 101854-060
Fmoc-L-(2,6-di-Me)Tyr-OH
Fmoc-L-(2,6-di-Me)Tyr-OH

Hazard: Irritant

Formula: C₂₆H₂₅NO₅
MDL Number: MFCD01632251
CAS Number: 206060-54-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR