Fmoc-D-thyroxine
Supplier: Matrix Scientific
042002-1G
042002-5G
101854-054EA
823.08
USD
101854-054
101854-056
Fmoc-D-thyroxine
Fmoc-D-thyroxine
Hazard: Irritant
Formula:
C₃₀H₂₁I₄NO₆ |
MDL Number:
MFCD02094440 CAS Number: 1217630-45-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...