Fmoc-DL-O-tyrosine

Supplier: Matrix Scientific

042000-1G 042000-5G
101854-046EA 1095.38 USD
101854-046 101854-048
Fmoc-DL-O-tyrosine
Fmoc-DL-O-tyrosine

Hazard: Irritant

Formula: C₂₄H₂₁NO₅
MW: 403.44 g/mol
MDL Number: MFCD02682300
CAS Number: 174879-27-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR