Fmoc-DL-m-tyrosine

Supplier: Matrix Scientific

041999-1G 041999-5G
101854-042EA 1095.38 USD
101854-042 101854-044
Fmoc-DL-m-tyrosine
Fmoc-DL-m-tyrosine

Hazard: Irritant

Formula: C₂₄H₂₁NO₅
MDL Number: MFCD02682298
CAS Number: 138775-49-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR