Fmoc-D-Homophe-OH

Supplier: Matrix Scientific

Synonyms: N-Fmoc-D-homophenylalanine, Fmoc-D-Homophe-OH, Fmoc-D-HoPhe-OH

041998-1G 041998-5G
101854-038EA 101.54 USD
101854-038 101854-040
Fmoc-D-Homophe-OH
Fmoc-D-Homophe-OH

Hazard: Irritant

Formula: C₂₅H₂₃NO₄
MW: 401.46 g/mol
MDL Number: MFCD00151917
CAS Number: 135944-09-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR