Fmoc-D-3,3-diphenylalanine

Supplier: Matrix Scientific

041997-1G 041997-5G
101854-034EA 436.92 USD
101854-034 101854-036
Fmoc-D-3,3-diphenylalanine
Fmoc-D-3,3-diphenylalanine

Hazard: Irritant

Formula: C₃₀H₂₅NO₄
MDL Number: MFCD00672338
CAS Number: 189937-46-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR